6-Bromosalicylaldehyde

Supplier: Apollo Scientific

Synonyms: 2-Bromo-6-hydroxybenzaldehyde

OR61135-1G OR61135-500MG OR61135-10G
APOSOR61135-1GEA 19.7 GBP
APOSOR61135-1G APOSOR61135-500MG APOSOR61135-10G
6-Bromosalicylaldehyde
6-Bromosalicylaldehyde
Formula: C₇H₅BrO₂
MW: 201.0200 g/mol
Melting Pt: 51…52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08234722
CAS Number: 22532-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR