N-(tert-Butoxycarbonyl)-2-(2-aminoethoxy)ethylamine
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
tert-Butyl (2-(2-aminoethoxy)ethyl)carbamate, N-Boc-2-(2-aminoethoxy)ethylamine, Amino-PEG2-NH-Boc, [2-(2-Aminoethoxy)ethyl]carbamic acid tert-Butyl ester, tert-Butyl [2-(2-Aminoethoxy)ethyl]carbamate
OR61058-1G
OR61058-250MG
OR61058-5G
APOSOR61058-1GEA
155
GBP
APOSOR61058-1G
APOSOR61058-250MG
APOSOR61058-5G
N-(tert-Butoxycarbonyl)-2-(2-aminoethoxy)ethylamine
N-(tert-Butoxycarbonyl)-2-(2-aminoethoxy)ethylamine
Formula:
C₉H₂₀N₂O₃ MW: 204,27 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12031501 CAS Number: 127828-22-2 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...