2-Bromo-5-chloropyrazine

Supplier: Apollo Scientific
Warning

OR60106-1G OR60106-5G
APOSOR60106-1GEA 48.6 GBP
APOSOR60106-1G APOSOR60106-5G
2-Bromo-5-chloropyrazine
2-Bromo-5-chloropyrazine
Formula: C₄H₂BrClN₂
MW: 193,4291 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08460074
CAS Number: 912773-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR