5-Nitro-2-picoline 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Methyl-5-nitropyridine
OR60065-1G
OR60065-25G
OR60065-5G
APOSOR60065-1GEA
44.6
GBP
APOSOR60065-1G
APOSOR60065-25G
APOSOR60065-5G
5-Nitro-2-picoline 95%
5-Nitro-2-picoline
Formula:
C₆H₆N₂O₂ MW: 138,12 g/mol Melting Pt: 107…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD04114179 CAS Number: 21203-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...