1H-Indazole-4-carboxylic acid

Supplier: Apollo Scientific
Warning

OR60061-1G OR60061-5G OR60061-10G
APOSOR60061-1GEA 25 GBP
APOSOR60061-1G APOSOR60061-5G APOSOR60061-10G
1H-Indazole-4-carboxylic acid
1H-Indazole-4-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03840644
CAS Number: 677306-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR