4-Borono-L-phenylalanine

Supplier: Apollo Scientific
Warning

Synonyms: H-Phe(4-B)-OH, L-4-(2-Amino-2-carboxyethyl)benzeneboronic acid, (S)-2-Amino-3-(4-boronophenyl)propanoic acid

OR60018-1G OR60018-250MG
APOSOR60018-1GEA 104 GBP
APOSOR60018-1G APOSOR60018-250MG
4-Borono-L-phenylalanine
4-Borono-L-phenylalanine
Formula: C₉H₁₂BNO₄
Melting Pt: 283…293 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01075172
CAS Number: 76410-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR