1-Amino-2-methoxyethane 98%

Supplier: Apollo Scientific
Danger

Synonyms: 2-Methoxyethylamine

OR59868-100G OR59868-25G
APOSOR59868-100GEA 197 GBP
APOSOR59868-100G APOSOR59868-25G
1-Amino-2-methoxyethane 98%
1-Amino-2-methoxyethane
Formula: C₃H₉NO
MW: 75,11 g/mol
Boiling Pt: 95 °C
Melting Pt: –82 °C
Density: 0,874 g/cm³ (25 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008180
CAS Number: 109-85-3
EINECS: 203-712-1
UN: 2733
ADR: 3+8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR