Ethyl-4-oxocyclohexanecarboxylate 98+%

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 4-oxocyclohexanecarboxylate, 4-(Ethoxycarbonyl)-1-cyclohexanone

OR5848-100G OR5848-25G OR5848-5G
APOSOR5848-100GEA 70.9 GBP
APOSOR5848-100G APOSOR5848-25G APOSOR5848-5G
Ethyl-4-oxocyclohexanecarboxylate 98+%
Ethyl 4-oxocyclohexanecarboxylate

Used in the preparation of dopamine agonists Biorg.Med.Chem.Lett. 1993,3,639

Formula: C₉H₁₄O₃
MW: 170,21 g/mol
Boiling Pt: 150…152 °C/40mmHg
Density: 1,068 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00013285
CAS Number: 17159-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR