5-Bromo-2-oxo-2,3-dihydropyridine-3-carbonitrile 98%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromo-2-hydroxynicotinonitrile

OR5766-1G OR5766-5G
APOSOR5766-1GEA 32.8 GBP
APOSOR5766-1G APOSOR5766-5G
5-Bromo-2-oxo-2,3-dihydropyridine-3-carbonitrile 98%
5-Bromo-2-oxo-2,3-dihydropyridine-3-carbonitrile
Formula: C₆H₃BrN₂O
MW: 199,0048 g/mol
Melting Pt: 219…223 °C
Storage Temperature: Ambient
MDL Number: MFCD07363806
CAS Number: 405224-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR