2-Bromo-6-chloropyridine 98%
Supplier: Apollo Scientific
|
Danger
|
OR5747-100G
OR5747-25G
APOSOR5747-100GEA
498
GBP
APOSOR5747-100G
APOSOR5747-25G
2-Bromo-6-chloropyridine 98%
2-Bromo-6-chloropyridine
Formula:
C₅H₃BrClN Melting Pt: 90…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00181262 CAS Number: 5140-72-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...