4-Bromo-N,N-diethylbenzenesulphonamide 97%

Supplier: Apollo Scientific
Warning

OR5700-1G OR5700-5G
APOSOR5700-1GEA 43.5 GBP
APOSOR5700-1G APOSOR5700-5G
4-Bromo-N,N-diethylbenzenesulphonamide 97%
4-Bromo-N,N-diethylbenzenesulphonamide
Formula: C₁₀H₁₄BrNO₂S
Storage Temperature: Ambient
MDL Number: MFCD00587586
CAS Number: 90944-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR