3'-Ethylacetophenone
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3'-Ethylacetophenone, m-Ethylacetophenone, 1-(3-Ethylphenyl)ethanone
OR5698-10G
OR5698-1G
OR5698-5G
APOSOR5698-10GEA
418
GBP
APOSOR5698-10G
APOSOR5698-1G
APOSOR5698-5G
3'-Ethylacetophenone
3'-Ethylacetophenone
Formula:
C₁₀H₁₂O MW: 148,2 g/mol Boiling Pt: 114…118 °C/15mmHg Density: 0,9912 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009660 CAS Number: 22699-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...