Methyl oxindole-6-carboxylate 98%

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 2-Oxoindoline-6-carboxylate, 2-Oxoindoline-6-carboxylic acid methyll ester

OR5623-1G OR5623-25G OR5623-5G
APOSOR5623-1GEA 46 GBP
APOSOR5623-1G APOSOR5623-25G APOSOR5623-5G
Methyl oxindole-6-carboxylate 98%
Methyl oxindole-6-carboxylate
Formula: C₁₀H₉NO₃
MW: 191,1800 g/mol
Melting Pt: 184…190 °C
Storage Temperature: Ambient
MDL Number: MFCD03095196
CAS Number: 14192-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR