(4-Bromo-2-nitrophenyl)acetic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Bromo-2-nitrophenylacetic acid

OR5592-1G OR5592-5G
APOSOR5592-1GEA 77.4 GBP
APOSOR5592-1G APOSOR5592-5G
(4-Bromo-2-nitrophenyl)acetic acid 97%
(4-Bromo-2-nitrophenyl)acetic acid
Formula: C₈H₆BrNO₄
MW: 260,0400 g/mol
Melting Pt: 156…160 °C
Storage Temperature: Ambient
MDL Number: MFCD05663950
CAS Number: 6127-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR