4-Nitrosalicylic acid 96%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Hydroxy-4-nitrobenzoic acid
OR55682-100G
OR55682-25G
OR55682-5G
APOSOR55682-100GEA
320
GBP
APOSOR55682-100G
APOSOR55682-25G
APOSOR55682-5G
4-Nitrosalicylic acid 96%
4-Nitrosalicylic acid
Formula:
C₇H₅NO₅ MW: 183.12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00071505 CAS Number: 619-19-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...