Piperidin-4-one hydrochloride 95%

Supplier: Apollo Scientific
Warning

OR55344-25G OR55344-100G
APOSOR55344-25GEA 28.9 GBP
APOSOR55344-25G APOSOR55344-100G
Piperidin-4-one hydrochloride 95%
Piperidin-4-one hydrochloride
Formula: C₅H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD00041019
CAS Number: 41979-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR