1-Dodecyne

Supplier: Apollo Scientific
Danger

OR5525-25G OR5525-5G
APOSOR5525-25GEA 314 GBP
APOSOR5525-25G APOSOR5525-5G
1-Dodecyne
1-Dodecyne
Formula: C₁₂H₂₂
Boiling Pt: 215 °C
Melting Pt: –19 °C
Density: 0,778 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008960
CAS Number: 765-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR