Chromane-2-carboxylic acid 95+%

Supplier: Apollo Scientific
Warning

OR55170-1G OR55170-5G
APOSOR55170-1GEA 51.2 GBP
APOSOR55170-1G APOSOR55170-5G
Chromane-2-carboxylic acid 95+%
Chromane-2-carboxylic acid
Formula: C₁₀H₁₀O₃
MW: 178,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044719
CAS Number: 51939-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR