5'-Bromo-m-terphenyl
Supplier: Apollo Scientific
OR55071-25G
OR55071-5G
APOSOR55071-25GEA
40
GBP
APOSOR55071-25G
APOSOR55071-5G
5'-Bromo-m-terphenyl
5'-Bromo-m-terphenyl
Formula:
C₁₈H₁₃Br Melting Pt: 111 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00196170 CAS Number: 103068-20-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...