5-Allyl-3-methoxysalicylaldehyde
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Allyl-2-hydroxy-3-methoxybenzaldehyde
OR55035-1G
OR55035-5G
APOSOR55035-1GEA
46
GBP
APOSOR55035-1G
APOSOR55035-5G
5-Allyl-3-methoxysalicylaldehyde
5-Allyl-3-methoxysalicylaldehyde
Formula:
C₁₁H₁₂O₃ Boiling Pt: 167 °C/9,75mmHg Melting Pt: 48 °C Storage Temperature: Ambient |
MDL Number:
MFCD06408022 CAS Number: 22934-51-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...