2,5-Dihydrofuran
Supplier: Apollo Scientific
|
Danger
|
OR5462-100G
OR5462-25G
APOSOR5462-100GEA
178
GBP
APOSOR5462-100G
APOSOR5462-25G
2,5-Dihydrofuran
2,5-Dihydrofuran
Formula:
C₄H₆O MW: 70,09 g/mol Boiling Pt: 66…67 °C Density: 0,927 g/cm³ (25 °C) Flash Pt: 1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003219 CAS Number: 1708-29-8 EINECS: 216-957-4 UN: 3271 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...