Bis(4-formylphenyl) ether 99%

Supplier: Apollo Scientific
Warning

Synonyms: 4-(4-Formylphenoxy)benzaldehyde, 4,4'-Diformyldiphenyl ether, 4,4'-Oxydibenzaldehyde

OR54445-1G OR54445-5G OR54445-10G
APOSOR54445-1GEA 49.8 GBP
APOSOR54445-1G APOSOR54445-5G APOSOR54445-10G
Bis(4-formylphenyl) ether 99%
Bis(4-formylphenyl) ether
Formula: C₁₄H₁₀O₃
Melting Pt: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD08276812
CAS Number: 2215-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR