7-Chloro-1H-pyrazolo[4,3-d]pyrimidine

Supplier: Apollo Scientific
Warning

OR54433-250MG OR54433-100MG
APOSOR54433-250MGEA 582 GBP
APOSOR54433-250MG APOSOR54433-100MG
7-Chloro-1H-pyrazolo[4,3-d]pyrimidine
7-Chloro-1H-pyrazolo[4,3-d]pyrimidine
Formula: C₅H₃ClN₄
Storage Temperature: Refrigerator
MDL Number: MFCD14706421
CAS Number: 923282-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR