1,5-Diazobicyclo[4.3.0]-5-nonene 98%

Supplier: Apollo Scientific
Danger

Synonyms: DBN, 2,3,4,6,7,8-Hexahydropyrrolo[1,2-a]pyrimidine, 1,5-Diazabicyclo[4.3.0]-5-nonene

OR5394-100G OR5394-1KG OR5394-250G
APOSOR5394-100GEA 91.8 GBP
APOSOR5394-100G APOSOR5394-1KG APOSOR5394-250G
1,5-Diazobicyclo[4.3.0]-5-nonene 98%
1,5-Diazobicyclo[4.3.0]-5-nonene
Formula: C₇H₁₂N₂
MW: 124,19 g/mol
Boiling Pt: 95…98 °C/7,5mmHg
Density: 1,005 g/cm³ (25 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00005554
CAS Number: 3001-72-7
EINECS: 221-087-3
UN: 3267
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR