3,5-Dihydroxybenzonitrile 98%

Supplier: Apollo Scientific

OR5346-1G OR5346-25G OR5346-5G
APOSOR5346-1GEA 197 GBP
APOSOR5346-1G APOSOR5346-25G APOSOR5346-5G
3,5-Dihydroxybenzonitrile 98%
3,5-Dihydroxybenzonitrile
Formula: C₇H₅NO₂
MW: 135.12 g/mol
Boiling Pt: 311
Melting Pt: 185…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00016453
CAS Number: 19179-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR