2-Mercaptothiazole 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Sulphanyl-1,3-thiazole

OR5341-1G OR5341-5G
APOSOR5341-1GEA 97.1 GBP
APOSOR5341-1G APOSOR5341-5G
2-Mercaptothiazole 97%
2-Mercaptothiazole
Formula: C₃H₃NS₂
MW: 117.19 g/mol
Melting Pt: 79…80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00022449
CAS Number: 82358-09-6
EINECS: 423-680-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR