1-Pentene 98%

Supplier: Apollo Scientific
Danger

Synonyms: alpha-n-Amylene, Propylethylene

OR5326-100G OR5326-25G
APOSOR5326-100GEA 197 GBP
APOSOR5326-100G APOSOR5326-25G
1-Pentene 98%
1-Pentene
Formula: C₅H₁₀
MW: 70,13 g/mol
Boiling Pt: 29…31 °C
Melting Pt: –138 °C
Density: 0,64 g/cm³ (25 °C)
Flash Pt: –50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003567
CAS Number: 109-67-1
UN: 1108
ADR: 3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR