Hexylamine

Supplier: Apollo Scientific
Danger

Synonyms: 1-Hexylamine, 1-Aminohexane

OR5310-1L OR5310-250ML
APOSOR5310-1LEA 257 GBP
APOSOR5310-1L APOSOR5310-250ML
Hexylamine
Hexylamine
Formula: H₃C(CH₂)₄CH₂NH₂
MW: 101,19 g/mol
Boiling Pt: 129…131 °C
Melting Pt: –19 °C
Density: 0,773 g/cm³ (25 °C)
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00008240
CAS Number: 111-26-2
EINECS: 203-851-8
UN: 2734
ADR: 8+3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR