Methyl-3-(4-methoxyphenyl)propanoate 98%

Supplier: Apollo Scientific
Warning

Synonyms: Methyl p-methoxyhydrocinnamate, Methyl 3-(4-methoxyphenyl)propanoate

OR53092-1G OR53092-5G
APOSOR53092-1GEA 19.7 GBP
APOSOR53092-1G APOSOR53092-5G
Methyl-3-(4-methoxyphenyl)propanoate 98%
Methyl-3-(4-methoxyphenyl)propanoate
Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 120 °C
Melting Pt: 37…41 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01923356
CAS Number: 15823-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR