Isatin 98%
Supplier: Apollo Scientific
Synonyms:
2,3-Indolinedione, Indoline-2,3-dione, 1H-Indole-2,3-dione
OR53066-100G
OR53066-2.5KG
OR53066-500G
APOSOR53066-100GEA
60.3
GBP
APOSOR53066-100G
APOSOR53066-2.5KG
APOSOR53066-500G
Isatin 98%
Isatin
Formula:
C₈H₅NO₂ MW: 147,13 g/mol Melting Pt: 199…200 °C Flash Pt: 220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005718 CAS Number: 91-56-5 EINECS: 202-077-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...