Methyl benzenesulphonate 98%

Supplier: Apollo Scientific
Danger

OR5306-100G OR5306-1KG OR5306-500G
APOSOR5306-100GEA 23.7 GBP
APOSOR5306-100G APOSOR5306-1KG APOSOR5306-500G
Methyl benzenesulphonate 98%
Methyl benzenesulphonate
Formula: C₇H₈O₃S
MW: 172,2 g/mol
Boiling Pt: 147…150 °C/13mmHg
Melting Pt: –4 °C
Density: 1,269 g/cm³ (25 °C)
Flash Pt: 143 °C
Storage Temperature: Ambient
MDL Number: MFCD00014737
CAS Number: 80-18-2
EINECS: 201-256-8
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR