Cyclopropanecarboxylic acid 98%
Supplier: Apollo Scientific
|
Danger
|
OR5295-1KG
OR5295-250G
OR5295-50G
APOSOR5295-1KGEA
129
GBP
APOSOR5295-1KG
APOSOR5295-250G
APOSOR5295-50G
Cyclopropanecarboxylic acid 98%
Cyclopropanecarboxylic acid
Formula:
C₄H₆O₂ MW: 86,09 g/mol Boiling Pt: 182…184 °C/760mmHg Melting Pt: 14…19 °C Density: 1,081 g/cm³ (25 °C) Flash Pt: 72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001287 CAS Number: 1759-53-1 EINECS: 217-162-5 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...