Methyl-5-nitrosalicylate 95%
Supplier: Apollo Scientific
Synonyms:
Methyl 2-hydroxy-5-nitrobenzoate
OR52922-100G
OR52922-25G
OR52922-500G
APOSOR52922-100GEA
237
GBP
APOSOR52922-100G
APOSOR52922-25G
APOSOR52922-500G
Methyl-5-nitrosalicylate 95%
Methyl 5-nitrosalicylate
Formula:
C₈H₇NO₅ MW: 197,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00024695 CAS Number: 17302-46-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...