Methyl-3,5-diaminobenzoate 99%

Supplier: Apollo Scientific

Synonyms: 3,5-Diaminobenzoic acid methyl ester, Methyl 3,5-diaminobenzoate

OR52894-10G OR52894-50G
APOSOR52894-10GEA 78.7 GBP
APOSOR52894-10G APOSOR52894-50G
Methyl-3,5-diaminobenzoate 99%
Methyl 3,5-diaminobenzoate
Formula: C₈H₁₀N₂O₂
MW: 166,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041433
CAS Number: 1949-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR