Fmoc-N-Me-Ile-OH 97%

Supplier: Apollo Scientific
Warning

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-isoleucine, Fmoc-N-Me-Ile-OH, N-Fmoc-N-methyl-L-isoleucine

OR52601-10G OR52601-25G OR52601-5G
APOSOR52601-10GEA 87.8 GBP
APOSOR52601-10G APOSOR52601-25G APOSOR52601-5G
Fmoc-N-Me-Ile-OH 97%
Fmoc-N-Me-Ile-OH
Formula: C₂₂H₂₅NO₄
MW: 367,445 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00153389
CAS Number: 138775-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR