N-Methylbiphenyl-2-amine 95%

Supplier: Apollo Scientific
Warning

OR52423-1G OR52423-250MG OR52423-5G OR52423-25G
APOSOR52423-1GEA 91.8 GBP
APOSOR52423-1G APOSOR52423-250MG APOSOR52423-5G APOSOR52423-25G
N-Methylbiphenyl-2-amine 95%
N-Methylbiphenyl-2-amine
Formula: C₁₃H₁₃N
MW: 183,2500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00179576
CAS Number: 14925-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR