Pentabromotoluene 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2,3,4,5,6-Pentabromotoluene

OR52367-25G OR52367-5G OR52367-1G
APOSOR52367-25GEA 78.7 GBP
APOSOR52367-25G APOSOR52367-5G APOSOR52367-1G
Pentabromotoluene 98%
Pentabromotoluene
Formula: C₇H₃Br₅
MW: 486,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000060
CAS Number: 87-83-2
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR