4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Pinacol vinylboronate, Vinylboronic Acid Pinacol Ester
OR52135-25G
OR52135-5G
OR52135-100G
APOSOR52135-25GEA
178
GBP
APOSOR52135-25G
APOSOR52135-5G
APOSOR52135-100G
4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane 95%
4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane
Formula:
C₈H₁₅BO₂ MW: 154,02 g/mol Boiling Pt: 35…40 °C/20mmHg Density: 0,908 g/cm³ (25 °C) Flash Pt: 34 °C Storage Temperature: Freezer |
MDL Number:
MFCD00192492 CAS Number: 75927-49-0 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...