5-Nitropicolinic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Nitro-2-pyridinecarboxylic acid, 5-Nitropyridine-2-carboxylic acid, 5-Nitropicolinic acid

OR52054-25G OR52054-5G
APOSOR52054-25GEA 172 GBP
APOSOR52054-25G APOSOR52054-5G
5-Nitropicolinic acid 98%
5-Nitropicolinic acid
Formula: C₆H₄N₂O₄
Melting Pt: 208…212 °C
Storage Temperature: Ambient
MDL Number: MFCD04114182
CAS Number: 30651-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR