3-Hydroxy-2-naphthoic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Hydroxy-naphthalene-2-carboxylic acid, 2-Hydroxy-3-naphthoic acid
OR51927-1KG
OR51927-250G
APOSOR51927-1KGEA
103
GBP
APOSOR51927-1KG
APOSOR51927-250G
3-Hydroxy-2-naphthoic acid 98%
3-Hydroxy-2-naphthoic acid
Formula:
C₁₁H₈O₃ MW: 188,18 g/mol Melting Pt: 218…221 °C Flash Pt: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004103 CAS Number: 92-70-6 EINECS: 202-180-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...