2-Amino-4-bromobenzaldehyde

Supplier: Apollo Scientific
Warning

OR51844-10G OR51844-1G OR51844-5G
APOSOR51844-10GEA 285 GBP
APOSOR51844-10G APOSOR51844-1G APOSOR51844-5G
2-Amino-4-bromobenzaldehyde
2-Amino-4-bromobenzaldehyde
Formula: C₇H₆BrNO
MW: 200,0326 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458822
CAS Number: 59278-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR