4-Isopropylbenzylamine 98%

Supplier: Apollo Scientific
Danger

Synonyms: (4-Isopropylphenyl)methanamine

OR51842-25G OR51842-5G
APOSOR51842-25GEA 289 GBP
APOSOR51842-25G APOSOR51842-5G
4-Isopropylbenzylamine 98%
4-Isopropylbenzylamine
Formula: C₁₀H₁₅N
MW: 149,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00025580
CAS Number: 4395-73-7
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR