(3,5-Dimethoxyphenyl)acetonitrile 96%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3,5-Dimethoxyphenylacetonitrile, 3,5-Dimethoxybenzyl cyanide, 2-(3,5-Dimethoxyphenyl)acetonitrile
OR5174-10G
OR5174-5G
APOSOR5174-10GEA
84
GBP
APOSOR5174-10G
APOSOR5174-5G
(3,5-Dimethoxyphenyl)acetonitrile 96%
(3,5-Dimethoxyphenyl)acetonitrile
Formula:
C₁₀H₁₁NO₂ MW: 177,2 g/mol Melting Pt: 52…55 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016395 CAS Number: 13388-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...