2-Aminobenzhydrol
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(2-Aminophenyl)(phenyl)methanol
OR5164-5G
OR5164-1G
APOSOR5164-5GEA
243
GBP
APOSOR5164-5G
APOSOR5164-1G
2-Aminobenzhydrol
2-Aminobenzhydrol
Formula:
C₁₃H₁₃NO Melting Pt: 114…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017097 CAS Number: 13209-38-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...