3-Phenylaniline 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Aminobiphenyl, 3-Biphenylylamine, [1,1'-Biphenyl]-3-amine, Biphenyl-3-ylamine

OR5148-5G OR5148-25G
APOSOR5148-5GEA 59.1 GBP
APOSOR5148-5G APOSOR5148-25G
3-Phenylaniline 98%
3-Phenylaniline
Formula: C₁₂H₁₁N
MW: 169,23 g/mol
Boiling Pt: 125 °C/1mmHg
Melting Pt: 28…33 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00047846
CAS Number: 2243-47-2
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR