3,6-Dibromo-9-(2-naphthalenyl)-9H-carbazole 99%

Supplier: Apollo Scientific

Synonyms: 3,6-Dibromo-9-(naphthalen-2-yl)-9H-carbazole

OR51020-1G OR51020-5G
APOSOR51020-1GEA 57.7 GBP
APOSOR51020-1G APOSOR51020-5G
3,6-Dibromo-9-(2-naphthalenyl)-9H-carbazole 99%
3,6-Dibromo-9-(2-naphthalenyl)-9H-carbazole
Formula: C₂₂H₁₃Br₂N
MW: 451,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29072769
CAS Number: 1221237-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR