2-Chlorobenzylamine 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
(2-Chlorophenyl)methanamine
OR5025-5G
OR5025-25G
APOSOR5025-5GEA
49.9
GBP
APOSOR5025-5G
APOSOR5025-25G
2-Chlorobenzylamine 98%
2-Chlorobenzylamine
Formula:
C₇H₈ClN MW: 141,6 g/mol Boiling Pt: 227 °C Density: 1,173 g/cm³ (25 °C) Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008108 CAS Number: 89-97-4 EINECS: 201-955-8 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...