3,4-Dimethoxyaniline 98%

Supplier: Apollo Scientific
Warning

OR4959-25G OR4959-100G OR4959-500G
APOSOR4959-25GEA 13.2 GBP
APOSOR4959-25G APOSOR4959-100G APOSOR4959-500G
3,4-Dimethoxyaniline 98%
3,4-Dimethoxyaniline
Formula: C₈H₁₁NO₂
MW: 153,18 g/mol
Boiling Pt: 174…176 °C/22mmHg
Melting Pt: 85…89 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008394
CAS Number: 6315-89-5
EINECS: 228-647-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR