Methyl-4-chloroacetoacetate

Supplier: Apollo Scientific
Danger

Synonyms: Methyl 4-chloro-3-oxobutanoate

OR4924-25G OR4924-100G
APOSOR4924-25GEA 23.7 GBP
APOSOR4924-25G APOSOR4924-100G
Methyl-4-chloroacetoacetate
Methyl 4-chloroacetoacetate
Formula: C₅H₇ClO₃
MW: 150,56 g/mol
Boiling Pt: 85 °C/4mmHg
Melting Pt: 14…16 °C
Density: 1,305 g/cm³ (25 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00000938
CAS Number: 32807-28-6
EINECS: 251-230-5
UN: 2922
ADR: 8+6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR