6-Oxo-1,6-dihydropyridine-3-carbaldehyde 98%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Hydroxynicotinaldehyde

OR4910-1G OR4910-5G OR4910-25G
APOSOR4910-1GEA 19.7 GBP
APOSOR4910-1G APOSOR4910-5G APOSOR4910-25G
6-Oxo-1,6-dihydropyridine-3-carbaldehyde 98%
6-Oxo-1,6-dihydropyridine-3-carbaldehyde
Formula: C₆H₅NO₂
Melting Pt: 222…226 °C
Storage Temperature: Ambient
MDL Number: MFCD03094941
CAS Number: 106984-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR